Assay Details
Target Gene Details
Entrez Gene ID: | 15170 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 6 |
Gene Aliases: |
70Z-SHP, Hcph, PTPTY-42, Ptp1C, SH-PTP1, SHP-1, hcp, me |
Location: |
Chr.6:124720707-124738709 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn6 | NM_001077705.2 | 16 | 2120 | NP_001071173.1 |
| NM_013545.3 | 16 | 2118 | NP_038573.2 | |
| AK132509.1 | 15 | 2208 | BAE21210.1 | |
| AK150509.1 | 16 | 2025 | BAE29622.1 | |
| AK151077.1 | 16 | 2039 | BAE30092.1 | |
| AK151623.1 | 16 | 2042 | ||
| AK159686.1 | 16 | 2078 | ||
| AK159833.1 | 16 | 2071 | ||
| AK167923.1 | 16 | 2072 | ||
| AK169804.1 | 16 | 2120 | BAE41378.1 | |
| BC012660.1 | 16 | 2063 | AAH12660.1 | |
| M68902.1 | 16 | 2070 | AAA37796.1 | |
| M90389.1 | 16 | 2020 | AAA40007.1 |
More Information
Set Membership: |
|

Genomic Map