Assay Details
Target Gene Details
Entrez Gene ID: | 320631 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 15 |
Gene Aliases: |
4930500I12Rik |
Location: |
Chr.7:120328486-120407687 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca15 | NM_177213.3 | 16 | 2435 | NP_796187.2 |
| AY243471.1 | 16 | 2435 | AAP73045.1 | |
| BC141350.1 | 16 | 2400 | ||
| BC141351.1 | 16 | 2400 |
More Information
Set Membership: |
|

Genomic Map