Assay Details
Target Gene Details
Entrez Gene ID: | 70420 |
Gene Name: | RIKEN cDNA 2610034B18 gene |
Gene Aliases: |
Arpin |
Location: |
Chr.7:79925359-79935264 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610034B18Rik | NM_027420.4 | 4 | 433 | NP_081696.1 |
| AK011654.1 | 4 | 491 | BAB27760.1 | |
| AK039350.1 | 4 | 441 | BAC30326.1 | |
| AK051261.1 | 4 | 434 | BAC34579.1 | |
| AK156013.1 | 4 | 501 | BAE33545.1 | |
| BC031379.1 | 4 | 432 | AAH31379.1 |

Genomic Map