Assay Details
Target Gene Details
Entrez Gene ID: | 19283 |
Gene Name: | protein tyrosine phosphatase, receptor type Z, polypeptide 1 |
Gene Aliases: |
AI850339, DSD-1-PG, PTPbeta, PTPzeta, Ptprz, Ptpz, RPTPz, Rptpbeta |
Location: |
Chr.6:22875458-23052916 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprz1 | NM_001081306.1 | 4 | 708 | NP_001074775.1 |
| AJ133130.1 | 4 | 488 | CAB41976.1 | |
| AK081698.1 | 4 | 709 | BAC38299.1 | |
| BC151071.1 | 4 | 433 | ||
| BC157965.1 | 4 | 433 |
More Information
Set Membership: |
|

Genomic Map