Assay Details
Target Gene Details
Entrez Gene ID: | 232491 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 1 |
Gene Aliases: |
- |
Location: |
Chr.6:142345625-142362630 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pyroxd1 | NM_183165.3 | NP_898988.2 | ||
| AK165677.1 | BAE38333.1 | |||
| BC027061.1 | AAH27061.1 |
Target Gene Details
Entrez Gene ID: | 19691 |
Gene Name: | RecQ protein-like |
Gene Aliases: |
RecQ1 |
Location: |
Chr.6:142350342-142387116 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Recql | NM_001204907.1 | NP_001191836.1 | ||
| AB017105.1 | BAA75086.1 | |||
| AK138325.1 | BAE23625.1 |
More Information
Set Membership: |
|

Genomic Map