Assay Details
Target Gene Details
Entrez Gene ID: | 28253 |
Gene Name: | solute carrier organic anion transporter family, member 1b2 |
Gene Aliases: |
7330442B20Rik, OATP-C, OATP2, Oatp1b2, Oatp4, Slc21a10, Slc21a6, lst-1, mlst-1 |
Location: |
Chr.6:141629452-141686646 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slco1b2 | NM_020495.1 | NP_065241.1 | ||
| AB031959.1 | BAB03272.1 | |||
| AF250912.1 | AAF74719.1 |
More Information
Set Membership: |
|

Genomic Map