Assay Details
Target Gene Details
Entrez Gene ID: | 232679 |
Gene Name: | zinc finger, C3HC type 1 |
Gene Aliases: |
1110054L24Rik, AU018540, AU019789, HSPC216, Nipa |
Location: |
Chr.6:30366384-30391040 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc3hc1 | NM_172735.2 | NP_766323.1 | ||
| AJ537495.1 | CAD61162.1 | |||
| AK088527.1 | BAC40404.1 | |||
| AK146228.1 | BAE26995.1 | |||
| AK151694.1 | BAE30618.1 | |||
| AK167465.1 | BAE39549.1 | |||
| AK167992.1 | BAE39982.1 | |||
| AK168071.1 | ||||
| BC037445.1 | AAH37445.1 | |||
| BC050141.1 | AAH50141.1 |

Genomic Map