Assay Details
Target Gene Details
Entrez Gene ID: | 243725 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 9A |
Gene Aliases: |
2810430P21Rik, 4930518N04Rik, 5330407E15, A230094E16Rik, BB181831, NRB, mKIAA1222, neurabin-I |
Location: |
Chr.6:4902747-5165661 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r9a | NM_181595.3 | NP_853626.1 | ||
AB091828.1 | BAC77245.1 | |||
AK029737.1 | ||||
AK138313.1 | BAE23620.1 | |||
AY308065.1 | AAQ77229.1 | |||
BC053748.1 | AAH53748.1 |
