Assay Details
Target Gene Details
Entrez Gene ID: | 21426 |
Gene Name: | transcription factor EC |
Gene Aliases: |
BB107417, Tcfec, bHLHe34 |
Location: |
Chr.6:16830837-16898462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfec | NM_031198.3 | NP_112475.1 | ||
AF077742.1 | AAD24426.1 | |||
AK137141.1 | BAE23248.1 | |||
AK156327.1 | BAE33676.1 | |||
AK159579.1 | BAE35199.1 | |||
BC098494.1 | AAH98494.1 |
