Assay Details
Target Gene Details
Entrez Gene ID: | 56463 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
AL033314, Tudor-SN |
Location: |
Chr.6:28480315-28935162 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snd1 | NM_019776.2 | NP_062750.2 | ||
AB021491.1 | BAA84944.1 | |||
AK034914.1 | ||||
AK088028.1 | BAC40105.1 | |||
AK133837.1 | BAE21874.1 | |||
AK159837.1 | BAE35415.1 | |||
AK161591.1 | BAE36479.1 | |||
AK162432.1 | BAE36913.1 | |||
AK167576.1 | BAE39639.1 | |||
AK201243.1 | ||||
BC007126.1 | AAH07126.3 |
