Assay Details
Target Gene Details
Entrez Gene ID: | 66349 |
Gene Name: | ATP5S-like |
Gene Aliases: |
2310004L02Rik, C030044E10Rik |
Location: |
Chr.7:25619414-25625550 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp5sl | NM_001290487.1 | 2 | 145 | NP_001277416.1 |
NM_025504.4 | 2 | 1070 | NP_079780.2 | |
AK002370.1 | 2 | 125 | BAB22048.1 | |
AK009150.1 | 2 | 128 | BAB26107.1 | |
AK039669.1 | 2 | 1071 | BAC30413.1 | |
BC024867.1 | 1 | 101 | AAH24867.1 |
