Assay Details
Target Gene Details
Entrez Gene ID: | 171504 |
Gene Name: | apolipoprotein B receptor |
Gene Aliases: |
Apob-48r, Apob48r |
Location: |
Chr.7:126584912-126589125 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apobr | NM_138310.1 | NP_612183.1 | ||
AF141335.1 | AAL54862.1 | |||
AK156422.1 | ||||
BC030718.1 | AAH30718.1 |
Target Gene Details
Entrez Gene ID: | 12752 |
Gene Name: | ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease) |
Gene Aliases: |
AI323623 |
Location: |
Chr.7:126571400-126585817 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cln3 | AK134036.1 |
