Assay Details
Target Gene Details
Entrez Gene ID: | 75736 |
Gene Name: | BCL2-like 12 (proline rich) |
Gene Aliases: |
2810475P17Rik, 5430429M05Rik, Bcl-L12, Bcl2-L12 |
Location: |
Chr.7:44991222-44997643 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl2l12 | NM_029410.3 | NP_083686.1 | ||
AK013411.1 | ||||
AK017362.1 | BAB30709.1 |
Target Gene Details
Entrez Gene ID: | 100502630 |
Gene Name: | predicted gene 15545 |
Gene Aliases: |
OTTMUSG00000023473 |
Location: |
Chr.7:44986900-44994601 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm15545 | NR_045266.1 | 2 | 932 | |
AK087248.1 | 2 | 910 |
