Assay Details
Target Gene Details
Entrez Gene ID: | 70333 |
Gene Name: | CD3E antigen, epsilon polypeptide associated protein |
Gene Aliases: |
1190028F09, 2610103M17Rik, ASE-1, Ase1, CAST, PAF49 |
Location: |
Chr.7:19356007-19359483 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd3eap | NM_145822.2 | NP_665821.1 | ||
| AK011811.1 |
More Information
Set Membership: |
|

Genomic Map