Assay Details
Target Gene Details
Entrez Gene ID: | 233801 |
Gene Name: | acyl-CoA synthetase medium-chain family member 4 |
Gene Aliases: |
O-MACS, OMACS |
Location: |
Chr.7:119690026-119714566 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsm4 | NM_178414.3 | 4 | 743 | NP_848501.1 |
| AK132127.1 | 4 | 744 | BAE20992.1 | |
| BC048390.1 | 4 | 679 | AAH48390.1 |
More Information
Set Membership: |
|

Genomic Map