Assay Details
Target Gene Details
Entrez Gene ID: | 11870 |
Gene Name: | ADP-ribosyltransferase 1 |
Gene Aliases: |
ADPRT, ARTC1, Yac-1 |
Location: |
Chr.7:102101706-102111758 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Art1 | NM_009710.4 | NP_033840.2 | ||
| AK003107.1 | BAB22570.1 | |||
| AK009284.1 | BAB26194.1 | |||
| BC063760.1 | AAH63760.1 | |||
| BC132492.1 | ||||
| BC132496.1 | ||||
| X95842.1 | CAA65099.1 |
More Information
Set Membership: |
|

Genomic Map