Assay Details
Target Gene Details
Entrez Gene ID: | 12577 |
Gene Name: | cyclin-dependent kinase inhibitor 1C (P57) |
Gene Aliases: |
AL024410, CDKI, Kip2, p57(kip2), p57Kip2 |
Location: |
Chr.7:143458339-143461072 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn1c | NM_001161624.1 | NP_001155096.1 | ||
| NM_009876.4 | NP_034006.3 | |||
| AK077844.1 | BAC37031.1 | |||
| AK132278.1 | ||||
| BC005412.1 | AAH05412.1 | |||
| CJ175313.1 | ||||
| U20553.1 | AAC52186.1 | |||
| U22399.1 | AAA85096.1 |
More Information
Set Membership: |
|

Genomic Map