Assay Details
Target Gene Details
Entrez Gene ID: | 26366 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 10 |
Gene Aliases: |
Bgp3, Cea10 |
Location: |
Chr.7:24777156-24784656 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam10 | NM_007675.4 | 4 | 952 | NP_031701.3 |
AK018529.1 | 4 | 932 | BAB31256.1 | |
BC003346.1 | 4 | 973 | AAH03346.1 | |
BE949028.1 | 1 | 109 | ||
L38422.1 | 4 | 916 | AAB64011.1 |
