Assay Details
Target Gene Details
Entrez Gene ID: | 12176 |
Gene Name: | BCL2/adenovirus E1B interacting protein 3 |
Gene Aliases: |
Nip3 |
Location: |
Chr.7:138890836-138909506 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bnip3 | NM_009760.4 | NP_033890.1 | ||
AF041054.1 | AAD02922.1 | |||
AK014223.1 | BAB29214.1 | |||
AK075943.1 | BAC36072.1 | |||
AK152610.1 | BAE31356.1 | |||
AK170865.1 | ||||
BC046603.1 | AAH46603.1 |
