Assay Details
Target Gene Details
Entrez Gene ID: | 29865 |
Gene Name: | calcium binding protein 5 |
Gene Aliases: |
- |
Location: |
Chr.7:13383295-13408880 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabp5 | NM_013877.3 | NP_038905.1 | ||
AB079605.1 | BAC15573.1 | |||
AF169161.1 | AAF25795.1 | |||
AK044544.1 | BAC31972.1 | |||
AK087416.1 | ||||
BC018253.1 | AAH18253.1 |
