Assay Details
Target Gene Details
Entrez Gene ID: | 81904 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 7 |
Gene Aliases: |
- |
Location: |
Chr.7:3332024-3368222 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng7 | NM_133189.3 | 1 | 100 | NP_573452.3 |
| AF361349.1 | 1 | 100 | AAL50044.1 | |
| AF458899.1 | 1 | 100 | AAM00596.1 | |
| AK032173.1 | 2 | 212 | BAC27742.1 | |
| BC141277.1 | 1 | 138 |
More Information
Set Membership: |
|

Genomic Map