Assay Details
Target Gene Details
Entrez Gene ID: | 260299 |
Gene Name: | cell adhesion molecule 4 |
Gene Aliases: |
Igdf4c, Igsf4c, Tsll2 |
Location: |
Chr.7:24482023-24504533 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadm4 | NM_153112.3 | 3 | 368 | NP_694752.1 |
| AK143204.1 | 3 | 369 | BAE25303.1 | |
| AY059394.1 | 3 | 292 | AAL29692.1 |

Genomic Map