Assay Details
Target Gene Details
Entrez Gene ID: | 71601 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 20 |
Gene Aliases: |
9130012D09Rik |
Location: |
Chr.7:19965352-19991113 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam20 | NM_027839.2 | NP_082115.2 | ||
AK018613.1 | BAB31307.1 | |||
AK033581.2 | BAE43285.1 | |||
BC049361.1 | AAH49361.1 |
