Assay Details
Target Gene Details
Entrez Gene ID: | 71601 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 20 |
Gene Aliases: |
9130012D09Rik |
Location: |
Chr.7:19965352-19991113 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam20 | NM_027839.2 | 2 | 166 | NP_082115.2 |
| AK018613.1 | 2 | 163 | BAB31307.1 | |
| AK033581.2 | 2 | 167 | BAE43285.1 | |
| BC049361.1 | 2 | 147 | AAH49361.1 |

Genomic Map