Assay Details
Target Gene Details
Entrez Gene ID: | 330657 |
Gene Name: | protease, serine 53 |
Gene Aliases: |
Pol3s, mFLJ00289 |
Location: |
Chr.7:127885444-127890970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss53 | NM_001081268.1 | NP_001074737.1 | ||
| AK220244.1 | BAD90169.1 | |||
| BC147509.1 | ||||
| BC151010.1 | ||||
| BC151017.1 | ||||
| BC172059.1 |
More Information
Set Membership: |
|

Genomic Map