Assay Details
Target Gene Details
Entrez Gene ID: | 75770 |
Gene Name: | BR serine/threonine kinase 2 |
Gene Aliases: |
4833424K13Rik, SAD-A, SADA |
Location: |
Chr.7:141949751-142004244 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brsk2 | NM_001009929.3 | NP_001009929.1 | ||
| NM_001009930.3 | NP_001009930.1 | |||
| NM_001276763.1 | NP_001263692.1 | |||
| NM_029426.2 | NP_083702.1 | |||
| AK158574.1 | ||||
| AK173268.1 | BAD32546.1 | |||
| AY533672.1 | AAT08447.1 | |||
| AY533673.1 | AAT08448.1 | |||
| AY533674.1 | AAT08449.1 | |||
| AY660739.1 | AAT74618.1 | |||
| BC056498.1 | AAH56498.1 | |||
| JF707771.1 | ||||
| JF707772.1 |
Target Gene Details
Entrez Gene ID: | 100526509 |
Gene Name: | microRNA 3104 |
Gene Aliases: |
mir-3104, mmu-mir-3104 |
Location: |
Chr.7:141992179-141992241 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir3104 | NR_037291.1 |
More Information
Set Membership: |
|

Genomic Map