Assay Details
Target Gene Details
Entrez Gene ID: | 210094 |
Gene Name: | IgLON family member 5 |
Gene Aliases: |
A230106M20Rik, Gm1431 |
Location: |
Chr.7:43472907-43490057 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iglon5 | NM_001164518.1 | NP_001157990.1 | ||
| AK039193.1 | BAC30273.1 | |||
| BC116421.1 | ||||
| BC116422.1 |
More Information
Set Membership: |
|

Genomic Map