Assay Details
Target Gene Details
Entrez Gene ID: | 67143 |
Gene Name: | IKAROS family zinc finger 5 |
Gene Aliases: |
2610034F18Rik, PEGASUS, Zfpn1a5 |
Location: |
Chr.7:131388649-131410506 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikzf5 | NM_175115.4 | NP_780324.2 | ||
| AK033716.1 | BAC28443.1 | |||
| AK088289.1 | BAC40260.1 | |||
| AK153883.1 | BAE32231.1 | |||
| AK169384.1 | BAE41131.1 | |||
| BC048183.1 | AAH48183.1 | |||
| BC058537.1 | AAH58537.1 | |||
| BC089455.1 | AAH89455.1 |
More Information
Set Membership: |
|

Genomic Map