Assay Details
Target Gene Details
Entrez Gene ID: | 19144 |
Gene Name: | kallikrein related-peptidase 6 |
Gene Aliases: |
AI849898, BSP, Bssp, Klk29, MSP, Prss18, Prss9, neurosin |
Location: |
Chr.7:43824544-43832027 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk6 | NM_001164696.1 | NP_001158168.1 | ||
| NM_001164697.1 | NP_001158169.1 | |||
| NM_001164698.1 | NP_001158170.1 | |||
| NM_011177.2 | NP_035307.2 | |||
| AB008928.1 | BAB55605.1 | |||
| AB015206.1 | BAA28895.1 | |||
| AK138218.1 | BAE23586.1 | |||
| BC031119.1 | AAH31119.1 | |||
| Y18723.1 | CAA77269.1 |
More Information
Set Membership: |
|

Genomic Map