Assay Details
Target Gene Details
Entrez Gene ID: | 23993 |
Gene Name: | kallikrein related-peptidase 7 (chymotryptic, stratum corneum) |
Gene Aliases: |
Prss6, SCCE |
Location: |
Chr.7:43811444-43816359 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk7 | NM_011872.2 | NP_036002.1 | ||
| AB008371.1 | BAB55604.1 | |||
| AF124299.1 | AAF01139.1 | |||
| AK029477.1 | BAC26467.1 | |||
| AK077406.1 | BAC36787.1 | |||
| BC027823.1 | AAH27823.1 |
More Information
Set Membership: |
|

Genomic Map