Assay Details
Target Gene Details
Entrez Gene ID: | 66085 |
Gene Name: | eukaryotic translation initiation factor 3, subunit F |
Gene Aliases: |
0610037M02Rik, AA409853, Eif3s5 |
Location: |
Chr.7:108934415-108941942 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3f | NM_025344.2 | NP_079620.2 | ||
| AK002778.1 | BAB22352.1 | |||
| AK003372.1 | ||||
| AK010333.1 | ||||
| AK088537.1 | BAC40412.1 | |||
| AK152037.1 | BAE30896.1 | |||
| AK190701.1 | ||||
| AK192917.1 | ||||
| AK197956.1 | ||||
| AK217928.1 | ||||
| BC070473.1 | AAH70473.1 | |||
| BC083190.1 | AAH83190.1 |
More Information
Set Membership: |
|

Genomic Map