Assay Details
Target Gene Details
Entrez Gene ID: | 73830 |
Gene Name: | eukaryotic translation initiation factor 3, subunit K |
Gene Aliases: |
1200009C21Rik, Eif3s12 |
Location: |
Chr.7:28971372-28981889 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3k | NM_001285942.1 | NP_001272871.1 | ||
| NM_028659.3 | NP_082935.1 | |||
| AK004664.1 | BAB23454.1 | |||
| BC027638.1 | AAH27638.1 | |||
| BC091749.1 | AAH91749.1 | |||
| CD802664.1 |
More Information
Set Membership: |
|

Genomic Map