Assay Details
Target Gene Details
Entrez Gene ID: | 16202 |
Gene Name: | integrin linked kinase |
Gene Aliases: |
AA511515, ESTM24 |
Location: |
Chr.7:105736590-105742925 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ilk | NM_001161724.1 | NP_001155196.1 | ||
| NM_010562.2 | NP_034692.2 | |||
| AK002344.1 | ||||
| AK011766.1 | ||||
| AK154207.1 | ||||
| BC003737.1 | AAH03737.1 | |||
| CA752166.1 | ||||
| U94479.1 | AAB94646.1 |
Target Gene Details
Entrez Gene ID: | 101867 |
Gene Name: | ribosomal RNA processing 8, methyltransferase, homolog (yeast) |
Gene Aliases: |
1500003O22Rik, 2900001K19Rik, AW538116 |
Location: |
Chr.7:105731730-105737385 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrp8 | NM_133951.1 | 1 | 135 | NP_598712.1 |
| AK154337.1 | 1 | 128 | BAE32523.1 |
More Information
Set Membership: |
|

Genomic Map