Assay Details
Target Gene Details
Entrez Gene ID: | 54131 |
Gene Name: | interferon regulatory factor 3 |
Gene Aliases: |
C920001K05Rik, IRF-3 |
Location: |
Chr.7:44997648-45002848 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf3 | NM_016849.4 | NP_058545.1 | ||
| NR_045611.1 | ||||
| AK013944.1 | ||||
| AK145977.1 | BAE26800.1 | |||
| AK146378.1 | BAE27126.1 | |||
| AK151750.1 | BAE30660.1 | |||
| AK152689.1 | BAE31420.1 | |||
| BC050882.1 | AAH50882.1 | |||
| EU545481.1 | ||||
| U75839.1 | AAB36924.1 | |||
| U75840.1 | AAB36925.1 |
More Information
Set Membership: |
|

Genomic Map