Assay Details
Target Gene Details
Entrez Gene ID: | 66065 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 14 |
Gene Aliases: |
0610039E24Rik, Dhrs10, retSDR3 |
Location: |
Chr.7:45554855-45567321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd17b14 | NM_025330.3 | NP_079606.3 | ||
| AK002831.1 | ||||
| AK010556.1 | BAB27025.1 | |||
| BC064042.1 | AAH64042.1 | |||
| BC099530.1 | AAH99530.1 |
More Information
Set Membership: |
|

Genomic Map