Assay Details
Target Gene Details
Entrez Gene ID: | 110826 |
Gene Name: | electron transferring flavoprotein, beta polypeptide |
Gene Aliases: |
0610009I16Rik, 2810441H06Rik |
Location: |
Chr.7:43444072-43457800 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etfb | NM_026695.3 | 6 | 784 | NP_080971.2 |
| NR_075104.1 | 6 | 783 | ||
| AI848359.1 | 1 | 137 | ||
| AK002407.1 | 6 | 726 | BAB22076.1 | |
| AK013284.1 | 6 | 748 | ||
| AK150293.1 | 6 | 727 | BAE29447.1 | |
| BC049237.1 | 6 | 713 | AAH49237.1 | |
| BC069877.1 | 6 | 743 | AAH69877.1 | |
| BE984117.1 | 1 | 117 |
More Information
Set Membership: |
|

Genomic Map