Assay Details
Target Gene Details
Entrez Gene ID: | 66245 |
Gene Name: | HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1 |
Gene Aliases: |
1500019G21Rik |
Location: |
Chr.7:4660515-4685121 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hspbp1 | NM_024172.3 | NP_077134.1 | ||
AF338351.1 | AAK11657.1 | |||
AK013263.1 | BAB28756.1 | |||
AK088469.1 | BAC40373.1 | |||
AK160128.1 | ||||
BC014758.1 | AAH14758.1 |
