Assay Details
Target Gene Details
Entrez Gene ID: | 56538 |
Gene Name: | kallikrein related-peptidase 11 |
Gene Aliases: |
2310015I08Rik, Prss20, TLSP |
Location: |
Chr.7:43774617-43779263 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk11 | NM_001177373.1 | NP_001170844.1 | ||
| NM_019974.2 | 1 | 68 | NP_064358.1 | |
| AB016226.1 | BAA88825.1 | |||
| AB016227.1 | BAA36955.1 | |||
| AK009360.1 | BAB26241.2 | |||
| AK009659.1 | ||||
| AK009720.1 | BAB26461.2 | |||
| BC148310.2 |
More Information
Set Membership: |
|

Genomic Map