Assay Details
Target Gene Details
Entrez Gene ID: | 23996 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, ATPase, 4 |
Gene Aliases: |
CIP21, MIP224 |
Location: |
Chr.7:28041702-28050092 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc4 | NM_011874.2 | NP_036004.2 | ||
| AK050677.1 | BAC34376.1 | |||
| AK077507.1 | BAC36835.1 | |||
| AK150989.1 | BAE30014.1 | |||
| AK153991.1 | BAE32301.1 | |||
| AK160644.1 | BAE35936.1 | |||
| AK160718.1 | BAE35967.1 | |||
| AK167041.1 | BAE39209.1 | |||
| AK167528.1 | BAE39598.1 | |||
| AK169224.1 | BAE40993.1 | |||
| AK169244.1 | ||||
| BC012708.1 | AAH12708.1 | |||
| BC092265.1 | AAH92265.1 | |||
| BC094063.1 | AAH94063.1 | |||
| L76223.1 | AAA88243.1 |
More Information
Set Membership: |
|

Genomic Map