Assay Details
Target Gene Details
Entrez Gene ID: | 20174 |
Gene Name: | RuvB-like protein 2 |
Gene Aliases: |
mp47, p47 |
Location: |
Chr.7:45421896-45435170 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ruvbl2 | NM_011304.3 | NP_035434.1 | ||
AB013912.1 | BAA76297.1 | |||
AK135404.1 | BAE22521.1 | |||
AK144156.1 | BAE25736.1 | |||
AK159112.1 | BAE34828.1 | |||
AK214338.1 | ||||
BC115809.1 | ||||
BC116693.1 |
