Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nomo1 | NM_153057.4 | 13 | 1569 | NP_694697.3 |
AK030584.1 | 13 | 1536 | ||
AK035957.1 | 13 | 1540 | ||
AK080754.1 | 13 | 1432 | ||
AK082495.1 | 13 | 1448 | BAC38508.1 | |
AK082963.1 | 13 | 1428 | BAC38713.1 | |
AK142338.1 | 13 | 1539 | ||
AK150107.1 | 13 | 1523 | ||
AK165643.1 | 13 | 1432 | ||
AK215705.1 | ||||
BC033923.1 | 2 | 153 | AAH33923.1 | |
BC072630.1 | 13 | 1515 | AAH72630.1 |
