Assay Details
Target Gene Details
Entrez Gene ID: | 214580 |
Gene Name: | phosphoseryl-tRNA kinase |
Gene Aliases: |
5430423O14Rik, 5730458D16Rik, Gm1099, R75284 |
Location: |
Chr.7:131371113-131392768 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pstk | NM_001039534.1 | 1 | 72 | NP_001034623.1 |
AK017332.1 | 1 | 62 | ||
AK077585.1 | 1 | 73 | BAC36878.1 | |
BC110381.1 | ||||
BC115503.2 | 1 | 73 |
