Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nomo1 | NM_153057.4 | NP_694697.3 | ||
AK030584.1 | ||||
AK035957.1 | ||||
AK080754.1 | ||||
AK082495.1 | BAC38508.1 | |||
AK082963.1 | BAC38713.1 | |||
AK142338.1 | ||||
AK150107.1 | ||||
AK165643.1 | ||||
AK166764.1 | BAE39002.1 | |||
AK190236.1 | ||||
AK200826.1 | ||||
AK201878.1 | ||||
AK201938.1 | ||||
AK202854.1 | ||||
AK205783.1 | ||||
AK211946.1 | ||||
AK213173.1 | ||||
BC033923.1 | AAH33923.1 | |||
BC072630.1 | AAH72630.1 |
