Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nomo1 | NM_153057.4 | 8 | 921 | NP_694697.3 |
AK030584.1 | 8 | 888 | ||
AK035957.1 | 8 | 891 | ||
AK080754.1 | 8 | 783 | ||
AK082495.1 | 8 | 800 | BAC38508.1 | |
AK082963.1 | 8 | 780 | BAC38713.1 | |
AK142338.1 | 8 | 891 | ||
AK150107.1 | 8 | 875 | ||
AK165643.1 | 8 | 782 | ||
AK199535.1 | ||||
AK200608.1 | 2 | 140 | ||
BC072630.1 | 8 | 867 | AAH72630.1 |
