Assay Details
Target Gene Details
Entrez Gene ID: | 233549 |
Gene Name: | monoacylglycerol O-acyltransferase 2 |
Gene Aliases: |
DGAT2L5, MGAT2, Mgat1l |
Location: |
Chr.7:99219084-99238611 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mogat2 | NM_177448.4 | 4 | 572 | NP_803231.1 |
| AK049560.1 | 4 | 536 | ||
| AK133854.1 | 4 | 580 | BAE21888.1 | |
| AY157609.1 | 4 | 529 | AAO23673.1 | |
| BC052831.1 | 4 | 576 | AAH52831.1 |
More Information
Set Membership: |
|

Genomic Map