Assay Details
Target Gene Details
Entrez Gene ID: | 791381 |
Gene Name: | predicted gene 15413 |
Gene Aliases: |
ENSMUSG00000053049, OTTMUSG00000021246 |
Location: |
Chr.7:96791433-96801549 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15413 | NR_045874.1 | |||
| AK077305.1 | BAC36740.1 |
Target Gene Details
Entrez Gene ID: | 23966 |
Gene Name: | teneurin transmembrane protein 4 |
Gene Aliases: |
Doc4, ELM2, Odz4, R75022, Ten-m4, l(7)-3Rn, l7Rn3, mKIAA1302 |
Location: |
Chr.7:96171237-96911093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tenm4 | NM_011858.3 | NP_035988.2 | ||
| AB025413.1 | BAA77399.1 | |||
| AF059485.1 | AAC31807.1 | |||
| AK039472.1 | ||||
| AK053790.1 | ||||
| AK147329.1 | BAE27851.1 | |||
| AK147579.1 | BAE28005.1 |
More Information
Set Membership: |
|

Genomic Map