Assay Details
Target Gene Details
Entrez Gene ID: | 67765 |
Gene Name: | RIKEN cDNA 5830432E09 gene |
Gene Aliases: |
- |
Location: |
Chr.7:135648457-135652314 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5830432E09Rik | NR_015548.1 | 1 | 114 | |
| AK017962.1 | 1 | 116 | ||
| AK020025.1 | 1 | 112 | ||
| AK088640.1 | 1 | 85 |
Target Gene Details
Entrez Gene ID: | 19267 |
Gene Name: | protein tyrosine phosphatase, receptor type, E |
Gene Aliases: |
PTPe, PTPepsilon, RPTPepsilon |
Location: |
Chr.7:135537645-135686294 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpre | NM_011212.3 | NP_035342.3 | ||
| AK004602.1 | ||||
| AK132455.1 | BAE21176.1 | |||
| AK154910.1 | BAE32920.1 | |||
| AK160947.1 | BAE36108.1 | |||
| BC024631.1 | ||||
| D83484.1 | BAA11927.1 | |||
| U35368.1 | AAC52281.1 | |||
| U36758.1 | AAC52331.1 | |||
| U40280.1 | AAB02190.1 | |||
| U62387.1 | AAB04553.1 |
More Information
Set Membership: |
|

Genomic Map