Assay Details
Target Gene Details
Entrez Gene ID: | 22342 |
Gene Name: | lin-7 homolog B (C. elegans) |
Gene Aliases: |
LIN-7B, MALS-2, Veli2 |
Location: |
Chr.7:45367891-45370564 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin7b | NM_011698.1 | NP_035828.1 | ||
| AF087694.1 | AAC78482.1 | |||
| AF173082.1 | AAD48501.1 | |||
| AK019299.1 | ||||
| BC031780.1 | AAH31780.1 |
More Information
Set Membership: |
|

Genomic Map