Assay Details
Target Gene Details
Entrez Gene ID: | 18631 |
Gene Name: | peroxisomal biogenesis factor 11 alpha |
Gene Aliases: |
PEX11alpha |
Location: |
Chr.7:79735957-79743126 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pex11a | NM_011068.1 | NP_035198.1 | ||
AF093669.1 | AAC78659.1 | |||
AK076776.1 | ||||
AK149387.1 | BAE28847.1 | |||
BC023439.1 | AAH23439.1 |
