Assay Details
Target Gene Details
| Entrez Gene ID: | 24030 | 
| Gene Name: | mitochondrial ribosomal protein S12 | 
| Gene Aliases: | AI327385, Rpms12, rps12 | 
| Location: | Chr.7:28739641-28741890 on Build GRCm38 | 
| Assay Gene Location: | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Mrps12 | AK146730.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic